Acetamid 99%, pure
Leverantör: Thermo Fisher Scientific
Varning
153631000 153635000 153630025
ACRO153631000EA 660 SEK
ACRO153631000 ACRO153635000 ACRO153630025
Acetamid 99%, pure
Acetamid
Formel: C₂H₅NO
MW: 59.07 g/mol
Kokpunkt: 221 °C (760 mmHg)
Smältpunkt: 79…81 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008023
CAS nummer: 60-35-5
EINECS: 200-473-5
Merck Index: 13,00044
Order Now
Specifikation testresultat
Appearance White Crystals
Infrared spectrum Conforms
Melting point 78 to 81 °C
GC ≥98.5 %