N,N-Dimetylformamid, extra dry over Molecular Sieve 99.8%, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
348430010 348430025 348431000 348435000
348430010.EA 2575 SEK
348430010. 348430025. 348431000. 348435000.
N,N-Dimetylformamid, extra dry over Molecular Sieve 99.8%, AcroSeal®
N,N-Dimetylformamid
Formel: HCON(CH₃)₂
Kokpunkt: 153 °C (760 mmHg)
Smältpunkt: –61 °C
Densitet: 0,945 g/cm³
Flampunkt: 58 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003284
CAS nummer: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.75 %
Acidity (CH3COOH) ≤0.01 %