N,N-Dimetylacetamid 99.5%, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC
378710010 378710025 378710200
ACRO378710010EA 995 SEK
ACRO378710010 ACRO378710025 ACRO378710200
N,N-Dimetylacetamid 99.5%, extra pure
N,N-Dimetylacetamid
Formel: CH₃CON(CH₃)₂
MW: 87,12 g/mol
Kokpunkt: 164…166 °C (760 mmHg)
Smältpunkt: –20 °C
Densitet: 0,937 g/cm³
Flampunkt: 70 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008686
CAS nummer: 127-19-5
EINECS: 204-826-4
ADR: 6.1,III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=833928cd423f317806bc60b3bc82ccdb74c2e20d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4370 to 1.4390 (20°C, 589 nm)
Water ≤0.1 %
GC ≥99.4 %
Color scale ≤10 APHA
Free acid (CH3COOH) ≤0.1 %