Valeryl chloride 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Pentanoyl chloride, Valeroyl chloride
169121000 169120010
ACRO169121000EA 835 SEK
ACRO169121000 ACRO169120010
Valeryl chloride 98%
Valeryl chloride
Formel: C₅H₉ClO
Kokpunkt: 125…127 °C (760 mmHg)
Smältpunkt: –110 °C
Densitet: 0,99 g/cm³
Flampunkt: 32 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000757
CAS nummer: 638-29-9
EINECS: 211-330-1
UN: 2502
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4230 (20°C, 589 nm)
GC ≥97.5 %
Titration RCOCl ≥97.5 %