Aluminium acetylacetonate
Leverantör: Thermo Scientific
Fara
Synonyms: Tris(2,4-pentanedionato-O,O')aluminium, Aluminium 2,4-pentanedionate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate
12542.22 12542.36
12542.22EA 1100 SEK
12542.22 12542.36
Aluminium acetylacetonate
Aluminium acetylacetonate
CAS No.: 13963-57-0 Molecular Formula: C15H21AlO6 Formula Weight: 324.31 Physical Form: Powder Density: 1.27 Melting Point: 192-194° Boiling Point: 314-316° Beilstein Reference: 4157942 UN No.: 3467 MDL No.: MFCD00000013
Formel: C₁₅H₂₁AlO₆
MW: 324,31 g/mol
Kokpunkt: 314…316 °C
Smältpunkt: 192…194 °C
Densitet: 1,27
Storage Temperature: Omgivande
MDL Number: MFCD00000013
CAS nummer: 13963-57-0
EINECS: 237-741-6
UN: 3467
ADR: 6.1,II