4,4-Dimethoxybutanone ≥90%, tech.
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4,4-Dimethoxybutan-2-one, 3-Oxobutyraldehyde dimethyl acetal, Acetoacetaldehyde-1,1-Dimethyl Acetal, Acetylacetaldehyde dimethyl acetal, 4,4-Dimethoxy-2-butanone
A12058.0B A12058.22 A12058.30
A12058.0BEA 8325 SEK
A12058.0B A12058.22 A12058.30
4,4-Dimethoxybutanone ≥90%, tech.
4,4-Dimethoxybutanone
Purity: 90% CAS No.: 5436-21-5 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 57°(134°F) Refractive Index: 1.4150 Melting Point: -83° Boiling Point: 170-172° Beilstein Reference: 1702372 UN No.: 1224 MDL No.: MFCD00008789
Formel: C₆H₁₂O₃
MW: 132.16 g/mol
Kokpunkt: 170…172 °C
Smältpunkt: –83 °C
Densitet: 0.996
Flampunkt: 57 °C (134 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008789
CAS nummer: 5436-21-5
EINECS: 226-605-1
UN: 1224
ADR: 3 III