Diphenyl carbonate 99%
Leverantör: Thermo Fisher Scientific
Varning
117230010 117231000 117230025 117230100
ACRO117230010EA 1325 SEK
ACRO117230010 ACRO117231000 ACRO117230025 ACRO117230100
Diphenyl carbonate 99%
Diphenyl carbonate
Formel: C₁₃H₁₀O₃
MW: 214.22 g/mol
Kokpunkt: 301…302 °C (760 mmHg)
Smältpunkt: 78…81 °C
Flampunkt: 168 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003037
CAS nummer: 102-09-0
EINECS: 203-005-8
UN: 3077
ADR: 9 III
Merck Index: 12,07433
Order Now
Specifikation testresultat
Appearance White Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 78 to 81 °C
GC ≥98.5 %
Residue after evaporation ≤0.003 %
Titration with NaOH ≥96.0 %
Dilution test passes test
Chloride (Cl) ≤0.001 %
Sulfite (SO3) passes test
Sulfate (SO4) ≤0.003 %
Ammonium (NH4) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤0.001 %
Free acid (CH3COOH) ≤0.4 %