3-Oxoglutaric acid 96%
Leverantör: Thermo Scientific
Varning
Synonyms: Acetone-1,3-dicarboxylic acid, 3-Ketoglutaric acid, 3-Oxopentanedioic acid, 1,3-Acetonedicarboxylic acid, β-Ketoglutaric acid
173151000 173155000
ACRO173151000EA 1500 SEK
ACRO173151000 ACRO173155000
3-Oxoglutaric acid 96%
3-Oxoglutaric acid
Formel: C₅H₆O₅
MW: 146,1 g/mol
Kokpunkt: 323 °C (1013 hPa)
Smältpunkt: 133 °C (dec.)
Densitet: 1,55 g/cm³ (20 °C)
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00002711
CAS nummer: 542-05-2
EINECS: 208-797-9
Merck Index: 13,00069
Order Now
Specifikation testresultat
Appearance White to beige Powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
Titration with NaOH ≥95 %
Sulfuric acid ≤2 %