Natriumpyruvat
Leverantör: Molekula
Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
19010543-500G 19010543-100G
MOLE19010543-500GEA 0 SEK
MOLE19010543-500G MOLE19010543-100G
Natriumpyruvat
Natriumpyruvat
Formel: CH₃COCOONa
MW: 110.04 g/mol
Smältpunkt: 220…230 °C
Densitet: 1.718 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002586
CAS nummer: 113-24-6
EINECS: 204-024-4