Natriumpyruvat, Millipore®
Leverantör: Merck
Varning
Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
M-Clarity™ quality level = MQ100
1.06619.0050 1.06619.0250 1.06619.1000
1.06619.0050EA 720 SEK
1.06619.0050 1.06619.0250 1.06619.1000
Natriumpyruvat, Millipore®
Natriumpyruvat
Formel: CH₃COCOONa
MW: 110.04 g/mol
Smältpunkt: 220…230 °C
Densitet: 1.718 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002586
CAS nummer: 113-24-6
EINECS: 204-024-4