Pyrodruvsyra 98%, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid
132140250 132141000 132145000 132140025
ACRO132140250EA 500 SEK
ACRO132140250 ACRO132141000 ACRO132145000 ACRO132140025
Pyrodruvsyra 98%, extra pure
Pyrodruvsyra
Formel: C₃H₄O₃
MW: 88,06 g/mol
Kokpunkt: 165 °C (760 mmHg)
Smältpunkt: 11,8 °C
Densitet: 1,25 g/cm³
Flampunkt: 82 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00002585
CAS nummer: 127-17-3
EINECS: 204-824-3
UN: 3265
ADR: 8,II
Merck Index: 12,08205
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4320 (20°C, 589 nm)
Specific gravity (20°C) 1.240 to 1.280
Titration with NaOH 97.5 to 102.5 %