Dietylpyrocarbonate
Leverantör: PanReac AppliChem
Varning
Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
A0881.0020 A0881.0100
A0881.0020EA 1625 SEK
A0881.0020 A0881.0100
Dietylpyrocarbonate
Dietylpyrocarbonate
Formel: O(COOC₂H₅)₂
MW: 162,14 g/mol
Kokpunkt: 93 °C (1013 hPa)
Densitet: 1,123 g/cm³ (20 °C)
Flampunkt: 69 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009106
CAS nummer: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088