Dietylpyrocarbonate 97%, ren
Leverantör: Thermo Scientific
Varning
Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
170250050 170250250 170251000 170255000
ACRO170250050EA 500 SEK
ACRO170250050 ACRO170250250 ACRO170251000 ACRO170255000
Dietylpyrocarbonate 97%, ren
Dietylpyrocarbonate
Formel: O(COOC₂H₅)₂
MW: 162,14 g/mol
Kokpunkt: 93…94 °C (18 mmHg)
Densitet: 1,1175 g/cm³
Flampunkt: 69 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009106
CAS nummer: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088
Order Now
Specifikation testresultat
Appearance Clear colorless to amber Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.4000 (20°C, 589 nm)
Specific gravity (20/4°C) 1.11 to 1.25
Titration excess NaOH ≥96.0 %