Etylacetylacetat 99+%, extra pure
Leverantör: Thermo Scientific
Varning
Synonyms: Acetoacetic acid ethyl ester, EAA
220401000 220400025 220405000
ACRO220401000EA 360 SEK
ACRO220401000 ACRO220400025 ACRO220405000
Etylacetylacetat 99+%, extra pure
Etylacetylacetat
Formel: C₆H₁₀O₃
MW: 130,14 g/mol
Kokpunkt: 180 °C (760 mmHg)
Smältpunkt: –45 °C
Densitet: 1,03 g/cm³
Flampunkt: 70 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009199
CAS nummer: 141-97-9
EINECS: 205-516-1
Merck Index: 13,03793
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4200 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.0 %
Color scale ≤10 APHA
Acidity (CH3COOH) ≤0.1 %