Succinonitrile 99+%

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: Butanedinitrile, Succinodinitrile

171181000 171185000
ACRO171181000EA 735 SEK
ACRO171181000 ACRO171185000
Succinonitrile 99+%
Succinonitrile
Formel: C₄H₄N₂
MW: 80.09 g/mol
Kokpunkt: 267
Smältpunkt: 57 °C
Flampunkt: 132
Storage Temperature: Kylskåp
MDL Number: MFCD00001949
CAS nummer: 110-61-2
EINECS: 203-783-9
Merck Index: 12,09041

Order Now

Specifikation testresultat

Appearance Colorless to white to light yellow Crystalline mass or waxy solid
Infrared spectrum Conforms
GC ≥96 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR