3-Hydroxypropiononitrile 99%
Leverantör: Thermo Fisher Scientific
Synonyms: Ethylene Cyanohydrin, 2-Cyanoethanol, 3-Hydroxypropionitrile, Hydracrylonitrile, β-Hydroxypropiononitrile
149390010 149390500 149392500
ACRO149390010EA 4625 SEK
ACRO149390010 ACRO149390500 ACRO149392500
3-Hydroxypropiononitrile 99%
3-Hydroxypropiononitrile
Formel: C₃H₅NO
Kokpunkt: 228 °C (1013 hPa)
Smältpunkt: –46 °C
Densitet: 1,04 g/cm³
Flampunkt: 128 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002826
CAS nummer: 109-78-4
EINECS: 203-704-8
Merck Index: 13,03828
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4246 to 1.4266 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -28° to -32° (20°C, 589 nm) (c=1 in DMF)