n-Butyronitril 99%
Leverantör: Thermo Scientific
Fara
Synonyms: Butannitril
108140025 108141000 108145000
ACRO108140025EA 4225 SEK
ACRO108140025 ACRO108141000 ACRO108145000
n-Butyronitril 99%
n-Butyronitril
Formel: C₄H₇N
MW: 69,11 g/mol
Kokpunkt: 115…117 °C (760 mmHg)
Smältpunkt: –112 °C
Densitet: 0,79 g/cm³
Flampunkt: 16 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001968
CAS nummer: 109-74-0
EINECS: 203-700-6
UN: 2411
ADR: 3,II
Merck Index: 13,01597
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3850 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤5 APHA
UV at 250 nm A: ≤0.005
Residue after evaporation ≤2 ppm
Acidity ≤0.0002 meq/g