2-Chloroethyl isocyanate 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Isocyanic acid 2-Chloroethyl ester
221190010 221190050 221190250
ACRO221190010EA 585 SEK
ACRO221190010 ACRO221190050 ACRO221190250
2-Chloroethyl isocyanate 98%
2-Chloroethyl isocyanate
Formel: C₃H₄ClNO
MW: 105,52 g/mol
Kokpunkt: 135 °C (1013 hPa)
Densitet: 1,237 g/cm³
Flampunkt: 56 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00002043
CAS nummer: 1943-83-5
EINECS: 217-734-4
UN: 3080
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4490 (20°C, 589 nm)
GC ≥97.5 %