1-Chloro-2-iodoethane 96% stabilised, liquid
Leverantör: Thermo Scientific
Varning
Stabilization: Stabilised with copper
L10742.03 L10742.06 L10742.14
L10742.03EA 920 SEK
L10742.03 L10742.06 L10742.14
1-Chloro-2-iodoethane 96% stabilised, liquid
1-Chloro-2-iodoethane
Purity: 96% CAS No.: 624-70-4 Molecular Formula: ClCH2CH2I Formula Weight: 190.41 Refractive Index: 1.5660 Boiling Point: 61-62°/30mm MDL No.: MFCD00041453
Formel: C₂H₄ClI
MW: 190,41 g/mol
Kokpunkt: 61…62 °C (30 mmHg)
Smältpunkt: –15,6 °C
Densitet: 2,088
Flampunkt: >100 °C (212 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00041453
CAS nummer: 624-70-4