Kloracetamid ≥98%

Leverantör: Thermo Scientific
Fara

Synonyms: 2-Chloroacetamide

A15238.30 A15238.0B A15238.0I
A15238.30EA 805 SEK
A15238.30 A15238.0B A15238.0I
Kloracetamid ≥98%
Kloracetamid

Purity: 98+% CAS No.: 79-07-2 Molecular Formula: ClCH2CONH2 Formula Weight: 93.51 Flash Point: 170°(338°F) Odour: Weak Boiling Point: 220° dec. Beilstein Reference: 878191 Merck Reference: 14,2110 UN No.: 2811 MDL No.: MFCD00008027

Formel: C₂H₄ClNO
MW: 93,51 g/mol
Kokpunkt: 220 °C (dec.)
Smältpunkt: 116…119 °C
Densitet: 1,6 g/cm³ (20 °C)
Flampunkt: 170 °C (338 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008027
CAS nummer: 79-07-2
EINECS: 201-174-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,2110

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR