Tridecanal 96% stabilised

Leverantör: Thermo Scientific
Varning

Synonyms: Tridecylaldehyde

A19722.06 A19722.14
A19722.06EA 1500 SEK
A19722.06 A19722.14
Tridecanal 96% stabilised
Tridecanal

Stabilized


Purity: 96% CAS No.: 10486-19-8 Molecular Formula: C13H26O Formula Weight: 198.35 Flash Point: 132°(269°F) Refractive Index: 1.4384 Odour: Characteristic Melting Point: 14-15° Boiling Point: 280° MDL No.: MFCD00007018

Formel: C₁₃H₂₆O
MW: 198,35 g/mol
Kokpunkt: 280 °C
Smältpunkt: 14…15 °C
Densitet: 0,83
Flampunkt: 132 °C (269 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007018
CAS nummer: 10486-19-8
EINECS: 234-004-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR