8-Bromo-1-octanol ≥95%
Leverantör: Thermo Fisher Scientific
Varning
H27628.03 H27628.06 H27628.14
H27628.03EA 1150 SEK
H27628.03 H27628.06 H27628.14
8-Bromo-1-octanol ≥95%
8-Bromo-1-octanol
Purity: 95% CAS No.: 50816-19-8 Molecular Formula: C8H17BrO Formula Weight: 209.13 Flash Point: 110°(230°F) Refractive Index: 1.4810 Boiling Point: 79-80°/0.07mm Beilstein Reference: 1900837 MDL No.: MFCD00010388
Formel: BrH₂C(CH₂)₆CH₂OH
Kokpunkt: 79…80 °C (0,07 mmHg)
Densitet: 1,220
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00010388
CAS nummer: 50816-19-8
EINECS: 256-785-7