6-Bromo-1-hexanol ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 6-Bromohexan-1-ol, Hexamethylene bromohydrin, 1-Bromo-6-hydroxyhexane
B21803.14 B21803.03 B21803.06
B21803.14EA 8950 SEK
B21803.14 B21803.03 B21803.06
6-Bromo-1-hexanol ≥97%
6-Bromo-1-hexanol
Purity: 97% CAS No.: 4286-55-9 Molecular Formula: Br(CH2)6OH Formula Weight: 181.08 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4820 Boiling Point: 105-106°/5mm MDL No.: MFCD00002983
Formel: BrH₂C(CH₂)₄CH₂OH
MW: 181,07 g/mol
Kokpunkt: 105…106 °C (5 mmHg)
Densitet: 1,384
Flampunkt: >110 °C (230 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00002983
CAS nummer: 4286-55-9