3-Bromo-1-propanol 92%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Bromopropan-1-ol, 1-Bromo-3-hydroxypropane
403200250 403200050
ACRO403200250EA 1900 SEK
ACRO403200250 ACRO403200050
3-Bromo-1-propanol 92%
3-Bromo-1-propanol
Formel: BrH₂CCH₂CH₂OH
MW: 138.99 g/mol
Kokpunkt: 62 °C (5 mmHg)
Densitet: 1.6 g/cm³
Flampunkt: 65
Storage Temperature: Kylskåp
MDL Number: MFCD00002942
CAS nummer: 627-18-9
EINECS: 210-986-6
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4900 (20°C, 589 nm)
GC ≥90 %