Bronopol ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Brom-2-nitropropan-1,3-diol
A11639.22 A11639.36 A11639.PG
A11639.22EA 3275 SEK
A11639.22 A11639.36 A11639.PG
Bronopol ≥98%
Bronopol
Purity: 98+% CAS No.: 52-51-7 Molecular Formula: C3H6BrNO4 Formula Weight: 199.99 pH: 5-7 Melting Point: ca 122-132° Beilstein Reference: 1705868 Merck Reference: 14,1447 UN No.: 3241 MDL No.: MFCD00007390
Formel: HOCH₂BrC(NO₂)CH₂OH
MW: 199,99 g/mol
Kokpunkt: 388 °C (1013 hPa)
Smältpunkt: ∼122…132 °C
Densitet: 1,91 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007390
CAS nummer: 52-51-7
EINECS: 200-143-0
UN: 3241
ADR: 4.1,III
Merck Index: 14,1447