Ethyl bromofluoroacetate ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Bromofluoroacetic ethyl ester, Ethylbromofluoroacetate
B21579.06 B21579.14
B21579.06EA 1200 SEK
B21579.06 B21579.14
Ethyl bromofluoroacetate ≥97%
Ethyl bromofluoroacetate
Purity: 97% CAS No.: 401-55-8 Molecular Formula: FBrCHCO2CH2CH3 Formula Weight: 184.99 Flash Point: 70°(158°F) Refractive Index: 1.4255 Boiling Point: 154° UN No.: 2810 MDL No.: MFCD00042095
Formel: BrFCHCOOCH₂CH₃
MW: 184,99 g/mol
Kokpunkt: 154 °C
Densitet: 1,540
Flampunkt: 70 °C (158 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00042095
CAS nummer: 401-55-8
EINECS: 206-928-4
UN: 2810
ADR: 6.1,III