6-Bromocaproic acid 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 6-Bromohexanoic acid
165670250 165671000 165675000
ACRO165670250EA 1050 SEK
ACRO165670250 ACRO165671000 ACRO165675000
6-Bromocaproic acid 98%
6-Bromocaproic acid
Formel: BrH₂C(CH₂)₄COOH
MW: 195,06 g/mol
Kokpunkt: 165…170 °C (20 mmHg)
Smältpunkt: 32…35 °C
Densitet: 1,4068 g/cm³ (30 °C)
Flampunkt: 67 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004422
CAS nummer: 4224-70-8
EINECS: 224-176-5
UN: 3261
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White to light orange Low melting crystals
Infrared spectrum Conforms
Melting point 32 to 35 °C
Titration with NaOH ≥97.5 %