4'-Bromoacetanilide 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Bromoacetanilide, N-(4-Bromophenyl)acetamide
171571000 171570250 171575000
ACRO171571000EA 1700 SEK
ACRO171571000 ACRO171570250 ACRO171575000
4'-Bromoacetanilide 98%
4'-Bromoacetanilide
Formel: H₃CC(O)NH(C₆H₄Br)
MW: 214,06 g/mol
Kokpunkt: 347 °C (1013 hPa)
Smältpunkt: 166…170 °C
Densitet: 1,717 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000092
CAS nummer: 103-88-8
EINECS: 203-154-9
Merck Index: 13,01383
Order Now
Specifikation testresultat
Appearance White to beige to light grey Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 166 to 170 °C
GC ≥97.5 %