3-Bromopropionyl chloride 95% tech.
Leverantör: Thermo Scientific
Fara
Synonyms: 3-Bromopropanoyl chloride
161990250 161991000
ACRO161990250EA 1350 SEK
ACRO161990250 ACRO161991000
3-Bromopropionyl chloride 95% tech.
3-Bromopropionyl chloride
Formel: BrH₂CCH₂COOCl
MW: 171,42 g/mol
Kokpunkt: 55…57 °C (17 mmHg)
Densitet: 1,7 g/cm³
Flampunkt: 79 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000746
CAS nummer: 15486-96-1
EINECS: 239-515-2
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4920 to 1.4970 (20°C, 589 nm)
GC ≥94 %