4'-Bromo-2'-chloroacetanilide ≥98%
Leverantör: Thermo Scientific
Varning
A18652.14 A18652.22
A18652.14EA 2075 SEK
A18652.14 A18652.22
4'-Bromo-2'-chloroacetanilide ≥98%
4'-Bromo-2'-chloroacetanilide
Purity: 98+% CAS No.: 3460-23-9 Molecular Formula: C8H7BrClNO Formula Weight: 248.51 Melting Point: 151-152° Beilstein Reference: 2719133
Formel: H₃CC(O)NH(C₆H₃BrCl)
MW: 248,51 g/mol
Smältpunkt: 151…152 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040852
CAS nummer: 3460-23-9