1,7-Octadiene ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Octa-1,7-diene
L07659.AE L07659.AP
L07659.AEEA 1075 SEK
L07659.AE L07659.AP
1,7-Octadiene ≥97%
1,7-Octadiene
Purity: 97% CAS No.: 3710-30-3 Molecular Formula: C8H14 Formula Weight: 110.20 Flash Point: 5°(41°F) Refractive Index: 1.4220 Odour: Strong Melting Point: -70° Boiling Point: 118-120° Beilstein Reference: 605288 UN No.: 2309 MDL No.: MFCD00008668
Formel: C₈H₁₄
MW: 110,2 g/mol
Kokpunkt: 118…120 °C
Smältpunkt: < –70 °C
Densitet: 0,735
Flampunkt: 5 °C (41 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008668
CAS nummer: 3710-30-3
EINECS: 223-054-9
UN: 2309
ADR: 3,II