3,4,7,8-Tetramethyl-1,10-phenanthroline 98+%
Leverantör: Thermo Scientific
Varning
172220010 172220050
ACRO172220010EA 1900 SEK
ACRO172220010 ACRO172220050
3,4,7,8-Tetramethyl-1,10-phenanthroline 98+%
3,4,7,8-Tetramethyl-1,10-phenanthroline
Formel: C₁₆H₁₆N₂
MW: 236,32 g/mol
Kokpunkt: 386 °C (1013 hPa)
Smältpunkt: 277…280 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004974
CAS nummer: 1660-93-1
EINECS: 216-762-4
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to light brown Powder
Infrared spectrum Conforms
Titration with HClO4 98.0 to 102.0 %
HPLC ≥97.5 %