1,10-Fenantrolin monohydrat, GR ACS analytical reagent, Supelco®
Leverantör: Merck
Fara
Synonyms: 1,10-Phenanthroline hydrate
M-Clarity™ quality level = MQ100
1.07225.0010 1.07225.0100
1.07225.0010EA 2000 SEK
1.07225.0010 1.07225.0100
1,10-Fenantrolin monohydrat, GR ACS analytical reagent, Supelco®
1,10-Fenantrolin monohydrat
Formel: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Smältpunkt: 93…94 °C
Densitet: 1.259 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149973
CAS nummer: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1 III
Merck Index: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Solubility 3.3 g/l (20 °C)
Melting Point 93 - 94 °C
Molar Mass 198.24 g/mol
Bulk Density 300 kg/m3
Hazard Class 6.1
Hazard Class Description Toxic materials