D-Glukonsyrans-1 ,5-lakton ≥99%
Leverantör: Thermo Fisher Scientific
Synonyms: GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone
A13105.0E A13105.30 A13105.36
A13105.0EEA 3725 SEK
A13105.0E A13105.30 A13105.36
D-Glukonsyrans-1 ,5-lakton ≥99%
D-Glukonsyrans-1 ,5-lakton
Purity: 99% CAS No.: 90-80-2 Molecular Formula: C6H10O6 Formula Weight: 178.14 Melting Point: 154-159° dec. Beilstein Reference: 83286 Merck Reference: 14,4457 Optical Rotation: +65° (c=4 in water) MDL No.: MFCD00006647
Formel: C₆H₁₀O₆
Kokpunkt: 385 °C (1013 hPa)
Smältpunkt: 154…159 °C (dec.)
Densitet: 1,61 g/cm³ (–5 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006647
CAS nummer: 90-80-2
EINECS: 202-016-5
Merck Index: 14,4457