(R)-(-)-2-Methylpiperazine 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (R)-2-Methylpiperazine
356270050 356270250
ACRO356270050EA 2800 SEK
ACRO356270050 ACRO356270250
(R)-(-)-2-Methylpiperazine 98%
(R)-(-)-2-Methylpiperazine
Formel: C₅H₁₂N₂
MW: 100.16 g/mol
Kokpunkt: 155…156 °C
Smältpunkt: 86…91 °C
Flampunkt: 65 °C
Storage Temperature: Omgivande
MDL Number: MFCD00192317
CAS nummer: 75336-86-6
UN: 2925
ADR: 4.1 II
Order Now
Specifikation testresultat
Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 86 to 91 °C
GC ≥97.5 %
Specific optical rotation -15° to -21° (20 °C, 589 nm) (c=1, toluene)