(S)-(-)-2-Phenyl-1-propanol ≥98%
Leverantör: Thermo Fisher Scientific
Varning
L13988.03 L13988.MD
L13988.03EA 4650 SEK
L13988.03 L13988.MD
(S)-(-)-2-Phenyl-1-propanol ≥98%
(S)-(-)-2-Phenyl-1-propanol
Purity: 98+% CAS No.: 37778-99-7 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 108°(226°F) Refractive Index: 1.5260 Boiling Point: 220° Optical Rotation: -18° (neat) MDL No.: MFCD00145249
Formel: C₉H₁₂O
Kokpunkt: 220 °C
Densitet: 0,975
Flampunkt: 108 °C (226 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00145249
CAS nummer: 37778-99-7