(S)-(-)-1-Phenylethanol 99%
Leverantör: Thermo Scientific
Varning
Synonyms: (S)-(-)-α-Methylbenzyl alcohol, (S)-(-)-sec-Phenethyl alcohol, (S)-1-Phenylethanol
217790010 217790050 217790250
ACRO217790010EA 670 SEK
ACRO217790010 ACRO217790050 ACRO217790250
(S)-(-)-1-Phenylethanol 99%
(S)-(-)-1-Phenylethanol
Formel: C₈H₁₀O
MW: 122,17 g/mol
Kokpunkt: 98 °C (20 mmHg)
Smältpunkt: 9…11 °C
Densitet: 1,01 g/cm³
Flampunkt: 85 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064264
CAS nummer: 1445-91-6
UN: 2937
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5260 to 1.5280 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -41° to -44° (20°C, 589 nm) (neat)