(S)-(-)-α-Metylbenzylamin 99+%, (99% ee)
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (S)-(-)-1-Fenyletylamin
151830250 151831000 151835000
ACRO151830250EA 820 SEK
ACRO151830250 ACRO151831000 ACRO151835000
(S)-(-)-α-Metylbenzylamin 99+%, (99% ee)
(S)-(-)-α-Metylbenzylamin
Formel: C₈H₁₁N
MW: 121.18 g/mol
Kokpunkt: 187…189 °C (760 mmHg)
Smältpunkt: –10 °C
Densitet: 0.94 g/cm³
Flampunkt: 71 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064406
CAS nummer: 2627-86-3
EINECS: 220-098-0
UN: 2735
ADR: 8 II
Merck Index: 12,06106
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5280 (20°C, 589 nm)
GC ≥99.0 %
Enantiomeric excess ≥98.5 % (GC)
Specific optical rotation -39° to -41° (20°C, 589 nm) (neat)
Titration with HClO4 ≥98.0 %