(R)-(+)-α-Metylbenzylamin ≥99%, ee 99%, ChiPros®
Leverantör: Thermo Scientific
Fara
Synonyms: (R)-(+)-1-Fenyletylamin
L19117.14 L19117.22 L19117.06
L19117.14EA 1125 SEK
L19117.14 L19117.22 L19117.06
(R)-(+)-α-Metylbenzylamin ≥99%, ee 99%, ChiPros®
(R)-(+)-α-Metylbenzylamin
Purity: 99+% CAS No.: 3886-69-9 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 71°(159°F) Refractive Index: 1.5260 Odour: Amine-like Melting Point: -10° Boiling Point: 187° Beilstein Reference: 2410916 Merck Reference: 14,6026 Optical Rotation: +30° (c=10 in ethanol) UN No.: 2735 MDL No.: MFCD00064405
Formel: C₈H₁₁N
MW: 121,18 g/mol
Kokpunkt: 187 °C
Smältpunkt: –10 °C
Densitet: 0,948
Flampunkt: 71 °C (159 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00064405
CAS nummer: 3886-69-9
EINECS: 223-423-4
UN: 2922
ADR: 8,II
Merck Index: 14,6026