(R)-(+)-α-Metylbenzylamin 99+%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (R)-(+)-1-Fenyletylamin
371462500 371460010 371460500
ACRO371462500EA 4475 SEK
ACRO371462500 ACRO371460010 ACRO371460500
(R)-(+)-α-Metylbenzylamin 99+%
(R)-(+)-α-Metylbenzylamin
produced by BASF AG
Formel: C₈H₁₁N
MW: 121.18 g/mol
Kokpunkt: 184…186 °C (760 mmHg)
Smältpunkt: –10 °C
Densitet: 0.95 g/cm³
Flampunkt: 75 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064405
CAS nummer: 3886-69-9
EINECS: 223-423-4
UN: 2922
ADR: 8 II
Merck Index: 12,06106
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5270 (20°C, 589 nm)
Water ≤0.5 %
GC ≥99.0 %
Specific optical rotation +39° to +41° (20°C, 589 nm) (neat)