(R)-(-)-4-Penten-2-ol 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (R)-(-)-1-Penten-4-ol, (R)-2-Hydroxypent-4-ene, (R)-(-)-Allyl methyl carbinol
B25659.03 B25659.06
B25659.03EA 2800 SEK
B25659.03 B25659.06
(R)-(-)-4-Penten-2-ol 95%
(R)-(-)-4-Penten-2-ol
Purity: 95% CAS No.: 64584-92-5 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 30°(86°F) Refractive Index: 1.4240 Boiling Point: 115-116° Optical Rotation: -5.2° (c=1 in chloroform) UN No.: 1987 MDL No.: MFCD03701536
Formel: C₅H₁₀O
MW: 86.13 g/mol
Kokpunkt: 115…116 °C
Densitet: 0.837
Flampunkt: 30 °C (86 °F)
Storage Temperature: Omgivande
MDL Number: MFCD03701536
CAS nummer: 64584-92-5
UN: 1987
ADR: 3 III