(S)-(+)-α-Methoxyphenylacetic acid ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: (S)-(+)-α-Methoxyphenylacetic acid, (S)-alpha-Methoxyphenylacetic Acid
L08748.03 L08748.MD
L08748.03EA 2375 SEK
L08748.03 L08748.MD
(S)-(+)-α-Methoxyphenylacetic acid ≥99%
(S)-(+)-α-Methoxyphenylacetic acid
Purity: 99% CAS No.: 26164-26-1 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 63-68° Boiling Point: 164-166°/18mm MDL No.: MFCD00064216
Formel: C₉H₁₀O₃
MW: 166,18 g/mol
Kokpunkt: 164…166 °C (18 mmHg)
Smältpunkt: 63…68 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064216
CAS nummer: 26164-26-1
EINECS: 247-492-5