L (-)-Tryptofan ≥99%
Leverantör: Thermo Scientific
Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
A10230.14 A10230.22 A10230.36
A10230.14EA 1125 SEK
A10230.14 A10230.22 A10230.36
L (-)-Tryptofan ≥99%
L (-)-Tryptofan
Purity: 99% CAS No.: 73-22-3 Molecular Formula: C11H12N2O2 Formula Weight: 204.23 Melting Point: 280° 285° Beilstein Reference: 86197 Merck Reference: 14,9797 Optical Rotation: -31° (c=1 in water) MDL No.: MFCD00064340
Formel: C₁₁H₁₂N₂O₂
MW: 204,23 g/mol
Smältpunkt: 280…285 °C
Densitet: 1,34 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064340
CAS nummer: 73-22-3
EINECS: 200-795-6
Merck Index: 14,9797