L (+)-Valin, Millipore®
Leverantör: Merck
Synonyms: H-Val-OH, Valine, (S)-(+)-Valine
M-Clarity™ quality level = MQ100
1.08495.0025 1.08495.0100 1.08495.1000 1.08495.9010
1.08495.0025EA 460 SEK
1.08495.0025 1.08495.0100 1.08495.1000 1.08495.9010
L (+)-Valin, Millipore®
L (+)-Valin
Formel: C₅H₁₁NO₂
MW: 117,15 g/mol
Kokpunkt: 217 °C (1013 hPa)
Smältpunkt: 86 °C
Densitet: 1,284 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064220
CAS nummer: 72-18-4
EINECS: 200-773-6
Merck Index: 13,09458
Order Now
Specifikation testresultat
Ammonium (NH4) 0.01% max.
Foreign Amino Acids 0.3% max.
Heavy Metals (as Pb) 0.001% max.
Melting Point 315°C