L(+)-Isoleucin, EMPROVE® ESSENTIAL Ph. Eur., USP suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH
1.05357.9010 1.05357.1000
1.05357.9010EA 0 SEK
1.05357.9010 1.05357.1000
L(+)-Isoleucin, EMPROVE® ESSENTIAL Ph. Eur., USP suitable for use as excipient, SAFC®
L(+)-Isoleucin
Formel: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17 g/mol
Smältpunkt: 279…280 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064222
CAS nummer: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179