(S)-(+)-2-Methylbutyric acid 98%
Leverantör: Thermo Fisher Scientific
Fara
224190010 224190050 224190250
ACRO224190010EA 1700 SEK
ACRO224190010 ACRO224190050 ACRO224190250
(S)-(+)-2-Methylbutyric acid 98%
(S)-(+)-2-Methylbutyric acid
Formel: C₅H₁₀O₂
Kokpunkt: 78 °C (15 mmHg)
Densitet: 0,938 g/cm³
Flampunkt: 73 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00063166
CAS nummer: 1730-91-2
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4070 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation +16° to +20° (20°C, 589 nm) (neat)