D (-)-Sorbitol ≥98%
Leverantör: Thermo Fisher Scientific
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
36404.36 36404.A3
36404.36EA 1025 SEK
36404.36 36404.A3
D (-)-Sorbitol ≥98%
D (-)-Sorbitol
Purity: 98% CAS No.: 50-70-4 Molecular Formula: C6H14O6 Formula Weight: 182.17 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 98-100° Solubility: Soluble in water, methanol, acetone, DMF Beilstein Reference: 1721899 Merck Reference: 14,8725 MDL No.: MFCD00004708
Formel: HOH₂C(CH(OH))₄CH₂OH
Kokpunkt: 295 °C (1013 hPa)
Smältpunkt: 98…100 °C
Densitet: 1,524 g/cm³ (20 °C)
Flampunkt: >100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004708
CAS nummer: 50-70-4
EINECS: 200-061-5
Merck Index: 14,8725
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM