(2R,3R)-(-)-2,3-Butanediol ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: D(-)-2,3-Butanediol, (2R,3R)-Butane-2,3-diol, (R,R)-(-)-2,3-Butylene glykol, (R,R)-(-)-2,3-Butanediol, (2R,3R)-(-)-2,3-Dihydroxy butane, (R,R)-(-)-2,3-Dihydroxy butane, (2R,3R)-(-)-2,3-Butylene glykol
L06586.03 L06586.06
L06586.03EA 1575 SEK
L06586.03 L06586.06
(2R,3R)-(-)-2,3-Butanediol ≥98%
(2R,3R)-(-)-2,3-Butanediol
Purity: 98% CAS No.: 24347-58-8 Molecular Formula: C4H10O2 Formula Weight: 90.12 Flash Point: 85°(185°F) Refractive Index: 1.4315 Melting Point: 16° Boiling Point: 179-180° Beilstein Reference: 4290593 Merck Reference: 14,1568 Optical Rotation: -13° (neat) MDL No.: MFCD00064267
Formel: C₄H₁₀O₂
MW: 90,12 g/mol
Kokpunkt: 179…180 °C
Smältpunkt: 16 °C
Densitet: 0,988
Flampunkt: 85 °C (185 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00064267
CAS nummer: 24347-58-8
EINECS: 246-186-9
Merck Index: 14,1568