(S)-(+)-2-Butanol 99%
Leverantör: Thermo Scientific
Varning
Synonyms: (S)-(+)-sec-Butyl alcohol, (S)-Butan-2-ol, (S)-(+)-sec-Butanol, (S)-(+)-Ethyl methyl carbinol, (S)-(+)-2-Hydroxy butane, (S)-(+)-Methyl ethyl carbinol
218030010 218030050
ACRO218030010EA 920 SEK
ACRO218030010 ACRO218030050
(S)-(+)-2-Butanol 99%
(S)-(+)-2-Butanol
Formel: C₄H₁₀O
MW: 74,12 g/mol
Kokpunkt: 99 °C (1013 hPa)
Smältpunkt: –114 °C
Densitet: 0,8 g/cm³
Flampunkt: 26 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064281
CAS nummer: 4221-99-2
EINECS: 224-168-1
UN: 1120
ADR: 3,III
Merck Index: 13,01540
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3980 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation +12° to +14° (20°C, 589 nm) (neat)