(R)-(-)-2-Amino-1-butanol 98%
Leverantör: Thermo Scientific
Fara
Synonyms: (R)-2-Aminobutan-1-ol, L(-)-2-Amino-1-butanol
244031000 244030250
ACRO244031000EA 2350 SEK
ACRO244031000 ACRO244030250
(R)-(-)-2-Amino-1-butanol 98%
(R)-(-)-2-Amino-1-butanol
Formel: C₄H₁₁NO
MW: 89,14 g/mol
Kokpunkt: 172…174 °C (760 mmHg)
Smältpunkt: –2 °C
Densitet: 0,947 g/cm³
Flampunkt: 90 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064419
CAS nummer: 5856-63-3
EINECS: 227-476-4
UN: 2735
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4495 to 1.4535 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation -6.5° to -10.5° (20°C, 589 nm) (neat)